Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(+)-Usnic acid

Supplier: Matrix Scientific
Synonyms: (9bR)-2,6-Diacetyl-7,9-dihydroxy-8,9b-dimethyldibenzo[b,d]furan-1,3(2H,9bH)-dione

064857-500MG
101928-514EA 183.08 USD InStock
101928-514
(+)-Usnic acid
(+)-Usnic acid
Hazard: Irritant


Formula: C₁₈H₁₆O₇
MW: 344.32 g/mol
MDL Number: MFCD00016878
CAS Number: 7562-61-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty