Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Pyridin-3-yl-1H-benzimidazole

Supplier: Matrix Scientific
Synonyms: 2-(Pyridin-3-yl)-1H-benzo[d]imidazole

066594-500MG
101931-966EA 484.62 USD InStock
101931-966
2-Pyridin-3-yl-1H-benzimidazole
2-Pyridin-3-yl-1H-benzimidazole
Hazard: Irritant


Formula: C₁₂H₉N₃
MW: 195.23 g/mol
MDL Number: MFCD00022665
CAS Number: 1137-67-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty