Would you like to log in again?
HyClone™ Classical Liquid Media: Medium 199, HyClone products (Cytiva)
Supplier: HyClone products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
HyClone™
SH30253.01 SH30253.FS SH30253.FS SH30253.01
18777-006EA 33.55 USD InStock
18777-006 CA18777-006 CA95053-164 95053-164
HyClone™ Classical Liquid Media: Medium 199, HyClone products (Cytiva)
Cell Culture Media
HyClone Medium 199 is manufactured using ISO 9001:2000-certified processes
  • Contains 5,6 mM glucose, and 0,7 mM L- glutamine
  • Available with or without phenol red and L-amino acids
  • With Earle’s salts for use in a CO2 incubator, or with Hank’s salts for use without CO₂
  • Hydrated using WFI quality water
  • 0,1 μm sterile filtered
  • Manufactured using ISO 9001:2000-certified processes
We can determine which (if any) ingredients in a medium formulation are of animal origin or, if they have come in contact with animal derived materials in manufacture.
High yield cultures begin with high quality media. In order to maintain these standards, all HyClone products as well as raw material components must pass strict Quality Control testing to confirm the appropriate level of quality. Each raw material has a specification document describing the product, the required grade or purity, the source, and the criteria for acceptance on incoming inspection.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty