Would you like to log in again?
Axygen® Pipette Filter Tip, Axygen®, Corning
Supplier: Corning
Axygen®
TF-1200-C-R-S TF-350-R-S TF-100 TF-200 TF-50 TF-300 TF-1000 TF-350 TF-400 TF-20
10011-890CS 2666.36 USD InStock
10011-890 10011-362 10011-882 10011-892 10011-886 10011-884 10011-348 80086-062 10011-888 10011-878
Axygen® Pipette Filter Tip, Axygen®, Corning
Pipette Tips
Axygen brand filter tips allow free passage of air while preventing passage of aerosols.
The filters contain no leachable additives.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty