Would you like to log in again?
Adapter, Tubing, Michel-Miller, Ace Glass Incorporated
Supplier: Ace Glass
5809-12
89052-146EA 96.16 USD InStock
89052-146
Adapter, Tubing, Michel-Miller, Ace Glass Incorporated
Fittings and Connectors
These stainless steel tubing adapters are for use with end fitting adapters to connect tubing to Michel-Miller columns.
  • Use with end fitting adapters
  • Threaded
  • Stainless steel
Thread is 1/4-inch-28 UNF. Extension opposite thread is 1.5mm O.D. (.060 inches).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty