Would you like to log in again?
Adapter, Bottom Drip, Internal Thread, Ace Glass Incorporated
Supplier: Ace Glass
5838-05 5838-54 5838-47 5838-10 5838-08 5838-43 5838-14 5838-51
89052-572EA 253.63 USD InStock
89052-572 89052-578 89052-566 89052-546 89052-530 89052-554 89052-520 89052-586
Adapter, Bottom Drip, Internal Thread, Ace Glass Incorporated
Fittings and Connectors
For use with chromatography columns, these bottom drip adapters have a shallow taper at the upper end that minimizes mixing below the packing support.
  • Shallow taper at upper end
  • Internal thread accepts tubing connectors
  • Available in PTFE or nylon and four different Ace-Thred sizes
Adapter has a 1/4-inch-28 UNF 2B internal thread that will accept tubing connectors. This adapter can also be used at the top of the column as an inlet adapter. Bore is 1mm.
Ordering information: Supplied with one FETFE o-ring and one polyethylene 100 micron filter disc packing support. Glass filter discs are also available. Contact your Avantor representative for further information. For optional filter disc remover, order 89052-532.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty