Would you like to log in again?
Adapter, End Fitting, Michel-Miller, Ace Glass Incorporated
Supplier: Ace Glass
5801-47 5801-48 5801-07 5801-57 5801-14 5801-53 5801-18 5801-46 5801-55 5801-22
89052-086EA 220.6 USD InStock
89052-086 89052-076 89052-082 89052-084 89052-080 89052-068 89052-078 89052-496 89052-498 89059-192
Adapter, End Fitting, Michel-Miller, Ace Glass Incorporated
Fittings and Connectors
This is a precision-made Michel-Miller replacement PTFE end fitting adapter.
  • Leak-tight seal without o-rings
  • Male thread is size designation matching column
  • Top female thread is 1/4-inch-28 UNF
  • Three options for retaining packing materials
Adapter is designed to make a leak-tight seal without the use of o-rings
There are three different options used to retain packing material: 1) paper filter discs; Porosity D (10-20 micron) glass filter discs; or PTFE screens. Choose the option that best meets your needs.
Simply tighten until white ring appears indicating a seal has been made, then secure lock nut; no need to over tighten. Male thread is size designation and matches thread size of column. Female thread at top is 1/4-inch-28 UNF with Heli-Coil insert to accept flanged end tubing fittings. Bore is 1.5mm (.060-inches).
Ordering information: Glass filter disc is supplied with appropriate adapters; adapters using paper filter discs or PTFE screens do NOT include those items. To retain packing material: Paper filter discs 89052-068, 89052-496, 89052-076, 89052-078, 89052-080; Porosity D (10-20 micron; 89052-860) glass filter discs 89052-082, 89052-084, 89052-086 or PTFE screens 89052-498, 89059-192.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty