Would you like to log in again?
Adjusta-Chrom, Recycling Column, Ace Glass Incorporated
Supplier: Ace Glass
5815-04 5815-30 5815-37 5815-08 5815-19 5815-20 5815-15 5815-03 5815-16 5815-33 5815-07 5815-26
89052-240EA 2002.47 USD InStock
89052-240 89052-242 89052-274 89052-230 89052-280 89052-248 89052-236 89052-238 89052-244 89052-232 89052-276 89052-246
Adjusta-Chrom, Recycling Column, Ace Glass Incorporated
Chromatography Columns
Adjustable bed chromatography column used for recycling, ascending or descending applications allowing for quick and easy adjustment of the PTFE plunger.
  • PTFE plunger available with PTFE or O-ring seals
  • Fabricated from borosilicate glass
The Porosity C (25 to 50 µm) glass disc gives rapid flow with a minimum of mixing. Pressure tight to 3.515 kg/cm2 (50 psi) at room temperature or slightly below.
Ordering information: Complete item includes column, plunger tip, appropriate extender(s) with screw cap(s), nylon bushing, PTFE tubing. These items are also available separately. Contact your Avantor representative for more information.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty