Would you like to log in again?
PhosphaGLO AP Substrate, SeraCare
Supplier: SeraCare
5430-0055 5430-0054
95059-356EA 260.46 USD InStock
95059-356 95059-358
PhosphaGLO AP Substrate, SeraCare
Assays Immunological Assays
PhosphaGLO AP Substrate is the chemiluminescent substrate of choice for easy, reliable detection of picogram protein levels of alkaline-phosphatase (AP) in Western blotting or ELISA
PhosphaGLO can replace other AP chemiluminescent substrates with little optimization required. It is compatible with film or chemiluminescent imager for flexibility in recording permanent results. When compared to other chemiluminescent AP substrates, PhosphaGLO offers significant cost savings per blot.
Product is a ready-to-use, 1-component substrate requiring no special blocking solutions. Long glow times up to five days allow for repeated exposures (Western Blot) or luminosity readings (ELISA). The protocol is easy-to-follow; simply add to microwell or blot and detect within minutes.PhosphaGLO Substrate is stable for two years at refrigerated temperatures; no need to be concerned about rapidly degrading substrates
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty