Would you like to log in again?
QuadMag Genie® Four-Position Magnetic Stirrers, Scientific Industries
Supplier: Scientific Industries
SI-5236
89202-326EA 1579.77 USD InStock
89202-326
QuadMag Genie® Four-Position Magnetic Stirrers, Scientific Industries
Stirrers Magnetic Stirrers
These four-position magnetic stirrers are available in an analog model or digital model
Choose the analog model, which is optimized for high speed, or choose the digital model for better speed control; this model offers digital display, control of speed, and is time-speed adjustable in 10rpm increments. Digital models also incorporate a timer from 1 to 99 minutes and a ramping feature that slowly increases speed to avoid splashing and improve spin bar control. Can be used in cold rooms and incubators.
Powerful, general purpose four-position magnetic stirrers feature "rare earth" magnets that stay coupled under the most demanding strirring conditions and offer exceptional magnetic force capable of stirring up to 2L for each position
Certifications: CE marked.
Ordering information: 230V models are supplied without a plug.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty