Would you like to log in again?
Union, UNF Female, Low Pressure, Ace Glass Incorporated
Supplier: Ace Glass
5854-40
89052-980EA 89.75 USD InStock
89052-980
Union, UNF Female, Low Pressure, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
Polyether ether ketone (PEEK) union for use with flanged tubing only.
  • PEEK union
  • Low pressure
  • UNF female to UNF female
Adapter is a 1/4-inch-28 UNF female to 1/4-inch-28 UNF female. For use with 1/8-inch O.D. tubing, this connector has a 1.5mm bore. Fittings are not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty