Would you like to log in again?
Total Hardness Test Strips, Hach
Supplier: Hach
2745250 2745250
97026-320EA 40.26 USD InStock
97026-320 CA97026-320
Total Hardness Test Strips, Hach
Environmental Test Kits
Water hardness is caused almost entirely by calcium and magnesium ions. Other di- and trivalent metals have a similar effect, but usually are not present in high enough concentration in potable waters to cause problems. Hardness increases soap consumption in laundries and causes scale in boilers.
Easy to use, disposable, and inexpensive.
A great way to obtain quick, quantitative answers in the field or in the lab.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty