Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium dodecyl sulfate (SDS) ≥99.0%

Supplier: IBI Scientific
Synonyms: Sodium dodecyl sulfateLauryl sulfate sodium saltSodium lauryl sulfateDodecyl sulfate sodium saltDodecyl sulphuric acid sodium saltLauryl sulphate sodium saltSodium lauryl sulphateDodecyl sulphate sodium saltDodecyl sulfuric acid sodium salt

IB07062
IB07062EA 165.13 USD InStock
IB07062
Sodium dodecyl sulfate (SDS) ≥99.0%
Sodium dodecyl sulphate (SDS)
SDS is an ionic detergent used to denature proteins in hybridization, nucleic purification, and electrophoresis buffer systems. SDS is also used to dissociate nucleic acid protein complexes in DNA extraction protocols, to disrupt cell membranes and to prepare prehybridization and/or hybridization solutions.

  • % of straight chain C12 SDS: >98
  • Chloride: <0.1%
  • Copper: <0.0005%
  • Heavy metals as Pb: ≤0.0005%
  • Phosphate: ≤0.0005%

Dnase-, Rnase, Protease- Free. Store at room temperature and protect from moisture.


Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Melting Pt: 204…207 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1,II
Merck Index: 13,08710
https://us-legacy.vwr.com/stibo/web/std.lang.all/54/91/735491.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty