Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Tissue Slice Chamber, BTX
Supplier: BTX
45-0501 750.75
95062-352EA 2558.26 USD InStock
95062-352 95062-354
Tissue Slice Chamber, BTX
Culture Dishes Cell Culture Dishes
This electrode chamber is designed for delicate and difficult tissue transfection
Ex-vivo electroporation is an efficient, effective method to introduce genes, drugs, or any number of molecules into ex-plant tissues. A common application is mouse brain slice for studying neuronal development. This specialty electrode makes transfection quick and simple, and is compatible with both the ECM® 830 and ECM® 2001 generators. The electrode is comprised of two parts, the glass petri dish electrode and the electrode wand. The glass petri dish contains a square-shaped platinum electrode chamber to secure the tissue sample. The wand incorporates an identical shaped platinum electrode, which is placed over the chamber to complete electroporation. This sandwich configuration ensures a homogeneous field of energy for optimal transfection.
Ordering information: Chamber Kit 95062-350 includes a 10x10mm tissue chamber, positive tissue electrode handle, and connector cables for both chamber and electrode. Tissue chamber 95062-352 includes a 10x10mm glass petri dish with platinum electrode chamber (connection cables must be ordered separetely).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty