Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Guanidinium thiocyanate ≥99.0%

Supplier: IBI Scientific
Synonyms: Guanidinium sulfocyanideGuanidine ThiocyanateGuanidinium sulphocyanideGTCGuanidinium rhodanide

IB05100
IB05100EA 251.25 USD InStock
IB05100
Guanidinium thiocyanate ≥99.0%
Guanidinium thiocyanate
Guanidine Thiocyanate is used in the isolation of RNA.

Dnase, Rnase, and Protease Free. Store at room temperature, tightly sealed, protect from moisture and light.


Formula: CH₅N₃·CHNS
MW: 118.16 g/mol
Melting Pt: 118…121 °C
MDL Number: MFCD00013027
CAS Number: 593-84-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty