Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Quest Micro Maxx Boingo Rockets

Supplier: QUEST AEROSPACE, INC.
Quest
5640 5634
470134-936EA 109.55 USD InStock
470134-936 470134-794
Quest Micro Maxx Boingo Rockets
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Tiny rockets, small launch area, big fun.

  • Skill level 1 (beginner)
  • Tumble recovery
  • Maximum altitude 37 m (120")
  • Requires Quest Micro Maxx rocket motors and initiators

Affordable Model Rocketry for groups in a micro size. With Quest's ARTF (Almost Ready-To-Fly) Micro Maxx™ kits, you'll enjoy the thrilling experience of model rocketry in a small yard. These micro-sized rockets require only a 50' Ø launch. Designed for backyard fun - you won't lose these micros the first time you launch them. Featuring the powerful Micro Maxx™ motor, maximum altitude is 120° and the tumble recovery method ensures that they land nearby.

Ordering information: Required but not included: One 9-volt battery for the launch controller.

Delivery information: All-Inclusive kit includes 12 Micro Maxx™ rocket kits, 12 MMX motors, 12 MMX-G2 initiators, 1 Micro Maxx™ launch pad and launch controller. Value 12-pack includes 12 Micro Maxx™ rocket kits (rockets only).

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty