Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Quest Planet Probe Model Rockets

Supplier: QUEST AEROSPACE, INC.
Quest
5476
470134-796EA 121.4 USD InStock
470134-796
Quest Planet Probe Model Rockets
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Ideal for schoolyards.

  • Skill level 1 (beginner)
  • Tumble recovery
  • Maximum altitude 76 m (250")
  • Requires 18 mm rocket motors (A or B recommended)

Fun-to-fly and easy-to-build. This cone shaped rocket is perfect for small flying areas. Great for 'drag races' or spot landing contests. No Wadding Required with gentle tumble recovery. This kit is great for kids. They can flip the pre-printed aeroshroud over during assembly for a plain white surface they can decorate on their own. Illustrates to consumers that a launch vehicle does not have to look like an arrow to fly like a rocket.

Quest skill level 1 beginner modeler kits may require a little measuring, gluing, sanding and painting. Assembly usually takes 45 to 90 minutes. Drying time for glue and painting could take up to 3 hours.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty