Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Quest Big Dog Advanced Enerjet Rockets

Supplier: QUEST AEROSPACE, INC.
Quest
5010
470134-804EA 63.25 USD InStock
470134-804
Quest Big Dog Advanced Enerjet Rockets
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Impressive flights, reasonable cost.

  • Skill level 3 (advanced)
  • Recovery with 16" nylon parachute
  • Flights up to 671 m (2,200")
  • Requires 24 mm rocket motors (E or F recommended)

Big Dog is big and basic. Standing over 32" tall, the kit features a blow molded plastic nose cone, laser-cut balsa fins and a rugged 24 mm diameter motor mount. Flights with an Enerjet F44-8W will go to almost 2,200. Then your Big Dog will recover gently on the big 16" nylon parachute. Kit includes big, easy-to-apply self-adhesive decals.

Enerjet skill Level 3 advanced modeler kits require moderate skills in model rocket construction and finishing. The assembly, finishing and glue drying time will take multiple sessions and up to a couple of days to complete the kit properly.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty