Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Quest Courier Model Rockets

Supplier: QUEST AEROSPACE, INC.
Quest
2011
470134-900EA 24.6 USD InStock
470134-900
Quest Courier Model Rockets
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Send and return a payload safely.

  • Skill level 2 (intermediate)
  • Recovery with dual 14" parachutes
  • Maximum altitude 274 m (900") without egg
  • Maximum altitude 137 m (450") with egg
  • Requires 18 mm rocket motors (A, B, or C recommended)

We can't think of many other ways that you can send an egg sailing 400 feet into the air and return it safely to Earth (and in time for breakfast). The specially designed nose capsule comes unpadded - requiring you to experiment with various methods of protecting the egg. The Courier model rocket has a traditional balsa fin design and features its own dual parachute recovery system.

Quest Level 2 experienced modeler kits require some basic beginner skills in model rocketry construction and finishing. Assembly, finishing and glue drying time can take a day or more.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty