Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Quest Payloader One Model Rockets

Supplier: QUEST AEROSPACE, INC.
Quest
1018
470134-902EA 20.3 USD InStock
470134-902
Quest Payloader One Model Rockets
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Payloader Plus Performance.

  • Skill level 1 (beginner)
  • Recovery with 14" parachute
  • Maximum altitude 366 m (1200")
  • Requires 18 mm rocket motors (A, B, or C recommended)

The Quest Payloader One model rocket kit is one of three Quest model rockets to use TTW Through-The-Wall balsa fin attachment. There are three precise pre-cut slots in the body tube that allow for perfect placement and location of the fins. Also features a semi-transparent payload compartment and parachute recovery system. The Payloader ONE model rocket mixes challenge and high performance.

Quest Skill Level 1 beginner modeler kits may require a little measuring, gluing, sanding and painting. Assembly usually takes 45 to 90 minutes. Drying time for glue and painting could take up to 3 hours.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty