Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Corning® Charcoal-Stripped FBS, Corning
Supplier: Corning
Corning®
35-072-CV 35-072-CV
97065-304EA 1419.11 USD InStock
CA97065-304 97065-304
Corning® Charcoal-Stripped FBS, Corning
Sera (Serum)
Charcoal-filtered (or “charcoal-stripped”) FBS is often used in immunoassays and hormone studies
Charcoal or charcoal-dextran removes lipophilic compounds (hormones, certain growth factors and cytokines) and virus particles.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty