Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nitric acid 64.8 - 65.2%, Supelco®

Supplier: MilliporeSigma
Danger

M-Clarity™ quality level = MQ200

NX0412-2 NX0412-2
EM-NX0412-2CS 1264 USD InStock
EM-NX0412-2 CANX0412-2
Nitric acid 64.8 - 65.2%, Supelco®
Nitric acid


Formula: HNO₃
MW: 63.01 g/mol
Boiling Pt: 120.5 °C (1013 hPa)
Melting Pt: –42 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II
Merck Index: 13,06608
Order Now
Specification Test Results
Arsenic (As) 0.005ppm max
Assay 64.8-65.2% by wt.
Chloride (Cl) 0.08ppm max
Fluorine (F) 1ppm max
Heavy metals (as Pb) 0.2ppm max
Iron (Fe) 0.2ppm max
Nonvolatile matter 5ppm max
Phosphate (PO4) 0.2ppm max
Sulfate (SO4) 0.08ppm max

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty