Would you like to log in again?
VWR® Glass Tube Inserts and Racks
Supplier: Avantor
V07-00002500 V07-HTGR1600
89237-554PK 43.91 USD InStock
89237-554 89237-552
VWR® Glass Tube Inserts and Racks
Racks Tube Racks
These round bottom borosilicate inserts with silicone mats are available in a range of volumes for a variety of sample preparation and sampling techniques.
  • Borosilicate glass fabrication
  • Wider opening allows for maximum sample removal
  • Silicone well covers
The wider opening of these inserts allow for pipetting deep within the insert for maximum sample removal. The racks are anodized aluminum and can be used in water baths, heaters and coolers and can then be transferred to other instruments. The round well silicone covers are available in both pre-cut or solid top for resealability. Inserts are sold in packs of 100 and covers are ten per pack. Racks are sold individually.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty