Would you like to log in again?
Acid Rain and the Environment: Acidity and Plant Growth
Supplier: ALDON CORP SE
IS9704
470136-736KT 87.35 USD InStock
470136-736
Acid Rain and the Environment: Acidity and Plant Growth
Educational Classroom Kits and Activities
Show How Acid Rain Affects Plants.
  • Real world hands on science
  • Ideal for biology, chemistry and earth science classrooms!
The problem of acid rain is quite often associated with its effects on aquatic systems. But it can also have devastating effects on terrestrial environments. In this activity, students will examine the detrimental effects of acidic conditions on plants grown under normal soil conditions, mildly acidic soil conditions, and very acidic soil conditions. Through physical observation, students will determine if the acidity has any impact on the growth of the plant.
Ordering information: Includes Hydrochloric Acid, Potting Soil, Transulcent Cups, Absorbent Wick Pad, Radish Seeds, and Teacher's Guide and Student Copymasters.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty