Would you like to log in again?
Leica DM300 Microscopes
Supplier: Leica
Leica®
13613384
470136-750EA 1706.35 USD InStock
470136-750
Leica DM300 Microscopes
Microscopes
Microscopes designed for the next generation of scientists.
  • Ergonomic body style
  • Light intensity control
  • Leica Limited Warranty
These Brightfield outfits include a DM300 Right Hand Stage Stand with a four-position nose piece, universal power supply, mechanical stage with slide holder, pre-centered and pre-focused Abbe condenser with adjustable iris and labels for matching objective magnifications, and LED illumination with continuous intensity control.
It also includes dust cover and user documents.
Ordering information: Power cord is sold separately.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty