Would you like to log in again?
Cylindrical Metal Spectroscope
Supplier: SCIENCE INTERACTIVE GROUP
614-0701
470136-842EA 41.1 USD InStock
470136-842
Cylindrical Metal Spectroscope
Educational Materials Physics Educational Materials Optics Learning Activities
View the Spectral Diffractions of Various Light Sources
This durable metal teaching spectroscope is an economical option for demonstrating the difference between incandescent lights and fluorescent lights right in your classroom. In addition to separating white light, it allows your students to see spectral diffractions of light sources including flames, solutions, colored glass, and excited gases. Plus, great colors are produced by its superior optics and a highly accurate diffraction grating. Includes spectroscope with focusing adjuster and instructions. Size: 100 mm x 25 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty