Would you like to log in again?
Combitips Advanced® Positive Displacement Pipette Tips for Eppendorf® Repeaters
Supplier: Eppendorf
Combitips® Advanced
0030089642 0030089413 0030089472 0030089685 0030089634 0030089480 0030089472 0030089448 0030089413 0030089618 0030089677 0030089421 0030089480 0030089677 0030089421 0030089669 0030089405 0030089464 0030089448 0030089430 0030089685 0030089626 0030089626 0030089693 0030089456 0030089634 0030089456 0030089642 0030089430 0030089693 0030089650 0030089464 0030089405 0030089669 0030089618 0030089650
89232-936CS 148.2 USD InStock
CA89232-980 CA89232-982 CA89232-940 CA89232-942 CA89232-944 CA89232-966 89232-936 CA89232-936 89232-934 89232-978 CA89232-938 89232-932 89232-976 89232-974 89232-930 89232-972 89232-970 CA89232-970 CA89232-972 CA89232-930 CA89232-974 CA89232-932 CA89232-976 89232-938 CA89232-978 CA89232-934 CA89232-946 CA89232-968 89232-946 89232-968 89232-944 89232-966 89232-942 89232-940 89232-982 89232-980
Combitips Advanced® Positive Displacement Pipette Tips for Eppendorf® Repeaters
Combitips Advanced® Positive Displacement pipette Tips are compatible with Eppendorf® Repeater® M4 Pipettor, Eppendorf® E3, Eppendorf® and E3X.
  • Individual color coding system facilitates the quick identification of the desired Combitips Advanced®
  • Elongated tips make it possible to empty all standard laboratory tubes completely
  • Funnel geometry prevents damage to gloves and ensures comfortable handling
  • Visibility of liquid volume is optimized by graduations
  • Nine volume sizes (0.1 to 50 ml) offer a maximum range of volumes
The Combitips Advanced® also function according to the positive displacement principle. Therefore, they always dispense the correct desired volume independently of the density of the liquid and its flow characteristics (e.g. increased vapor pressure or increased viscosity). Working with radioactive, aggressive, or toxic materials also becomes safer due to the hermetically sealed piston preventing aerosol contamination. After dispensing with the Repeater, simply eject and dispose of the Combitips Advanced® tips.
Tips have been completely redesigned and optimized so that they are capable of meeting the increased requirements of a modern laboratory. Combitips Advanced® are the ideal system components for all Repeaters.
Increments:
1 µl:0.1 µl
2 µl:0.2 µl
5 µl:0.5 µl
10 µl:1 µl
25 µl:2.5 µl
50 µl:5 µl
100 µl:10 µl
250 µl:25 µl
500 µl:50 µl
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty