Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diiodofluorescein 0.5% (w/v) in dilute alcohol USP Test Solution (TS)

Supplier: Ricca Chemical
Danger

R2554000-120A
RC2554-4EA 67.69 USD InStock
RC2554-4
Diiodofluorescein 0.5% (w/v) in dilute alcohol USP Test Solution (TS)
Diiodofluorescein


Formula: C₂₀H₁₀I₂O₅
MW: 584.11 g/mol
Boiling Pt: ~ 93 °C
Density: 0.8741 g/cm³
Flash Pt: ~ 23 °C
Storage Temperature: Ambient
MDL Number: MFCD00036461
CAS Number: 31395-16-1
UN: 1987
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear, orange liquid
Sensitivity Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty