Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfuric acid solution alcoholic solution for phosphorus determination

Supplier: Ricca Chemical

R0350000-1A
RC35032EA 130.42 USD InStock
RC35032
Sulfuric acid solution alcoholic solution for phosphorus determination
Sulfuric acid


Formula: H₂SO₄
MW: 98.08 g/mol
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 1830
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear, colorless, alcohol odor

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty