Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phosphotungstic acid USP Test Solution (TS)

Supplier: Ricca Chemical

R5859600-500A
RC5859.6-16EA 119.39 USD InStock
RC5859.6-16
Phosphotungstic acid USP Test Solution (TS)
Phosphotungstic acid


Formula: H₃O₄₀PW₁₂
MW: 2880.05 g/mol
CAS Number: 1343-93-7
UN: 1759
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear, colorless, odorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty