Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium carbonate 7% (w/v) in aqueous solution USP Test Solution (TS)

Supplier: Ricca Chemical
Danger
Synonyms: Carbonic acid potassium saltCarbonic acid, dipotassium salt

R5882500-500A
RC5882.5-16EA 139.7 USD InStock
RC5882.5-16
Potassium carbonate 7% (w/v) in aqueous solution USP Test Solution (TS)
Potassium carbonate


Formula: K₂CO₃
MW: 138.21 g/mol
Boiling Pt: ~ 100 °C
Density: 1.0604 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011382
CAS Number: 584-08-7
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/98/16640598.jpg
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 7.00 ± 0.02 % (w/v)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty