Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hydroxide 6.50% (w/v) in aqueous solution USP Test Solution (TS)

Supplier: Ricca Chemical
Synonyms: Potassium hydrateCaustic potash

R6125000-500A
RC6125-16EA 51.74 USD InStock
RC6125-16
Potassium hydroxide 6.50% (w/v) in aqueous solution USP Test Solution (TS)
Potassium hydroxide


Formula: KOH
MW: 56.11 g/mol
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1813
ADR: 8,II
Order Now
Specification Test Results
% (w/v) (traceable to NIST SRM 723) 6.50 ± 0.01 % (w/v)
Appearance Clear, colorless, odorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty