Would you like to log in again?
Rochelle Salt Stabilizer for Ammonia Nitrogen Determination, RICCA Chemical Company
Supplier: RICCA Chemical Company
R6640000-500A
RC664016EA 114.18 USD InStock
RC664016
Rochelle Salt Stabilizer for Ammonia Nitrogen Determination, RICCA Chemical Company
Environmental Test Kits
Reagent fo the APHA Nesslerization Method (direct and following distillation) for nitrogen (ammonia).
  • Ammonia (NH3): No detectable Ammonia
  • Appearance: Clear, colorless, odorless
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty