Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium carbonate 0.0454 N

Supplier: Ricca Chemical
Synonyms: Na-XCarbonic acid sodium saltDisodium carbonateSodaBisodium carbonateCarbonic acid, disodium saltSoda ash

R7180000-1A
RC718032EA 83.09 USD InStock
RC718032
Sodium carbonate 0.0454 N
Sodium carbonate
1 ml = 1 mg CO₂


Formula: Na₂CO₃
MW: 105.99 g/mol
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0025 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/89/16640589.jpg
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 0.04540 ± 0.00001 N at 20.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty