Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Disodium-L(+)-tartrate 11.5% (w/v) USP Test Solution (TS)

Supplier: Ricca Chemical

R7852000-500A
RC7852-16EA 95.81 USD InStock
RC7852-16
Disodium-L(+)-tartrate 11.5% (w/v) USP Test Solution (TS)
Disodium-L(+)-tartrate solutions


Formula: NaOOC(CHOH)₂COONa·2H₂O
MW: 230.08 g/mol
Boiling Pt: ~ 100 °C
Density: 1.0656 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00150035
CAS Number: 6106-24-7
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Refractive Index 11.5 ± 0.5 % (w/v)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty