Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lanthanum 5% in nitric acid 5% releasing agent

Supplier: Ricca Chemical

RMLA50KN-50N
RCMMLA50KN-50EA 685.27 USD InStock
RCMMLA50KN-50
Lanthanum 5% in nitric acid 5% releasing agent
Lanthanum


Formula: La
MW: 138.91 g/mol
MDL Number: MFCD00011066
CAS Number: 7439-91-0
UN: 3208
ADR: 4.3,II
Order Now
Specification Test Results
Appearance Clear, colorless liquid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty