Would you like to log in again?
Multi-Element Standard for ICP, Trace Metals II for Water, Ricca Chemical Company
Supplier: RICCA Chemical Company
RTRACE20-100B
RCRTRACE2-100EA 212.06 USD InStock
RCRTRACE2-100
Multi-Element Standard for ICP, Trace Metals II for Water, Ricca Chemical Company
Standards Cationic Standards Multi-Element Standards
This is a multielement solution that was prepared volumetrically to contain the elements and concentrations stated below
The uncertainty associated with the certified values is ± 0.5% relative. The concentrations are confirmed by ICP or ICP-MS
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty