Would you like to log in again?
Axygen® AxyPrep™ Mag Plasmid Kit, Corning
Supplier: Corning
Axygen® AxyPrep™
MAG-P-10
10159-702CS 318.55 USD InStock
10159-702
Axygen® AxyPrep™ Mag Plasmid Kit, Corning
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The AxyPrep™ Mag Plasmid purification kit utilizes a magnetic bead technology for high-throughput purification of plasmid DNA from E.coli cells.
AxyPrep™ Mag Plasmid kit can also be used with fosmid and BAC vector-based constructs ranging from 5kb to 150kb.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty