Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Orthophosphoric acid 1.00 N

Supplier: Ricca Chemical
Synonyms: Phosphoric acid

R5842500500A
RCR5842500500AEA 306.96 USD InStock
RCR5842500500A
Orthophosphoric acid 1.00 N
Orthophosphoric acid


Formula: H₃PO₄
MW: 98 g/mol
MDL Number: MFCD00011340
CAS Number: 7664-38-2
UN: 1805
ADR: 8,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty