Synonyms:
5-Amino-3-pyridinecarboxylic acid
A2000-5G
A2000-25G
TCA2000-5GEA
38.57
USD
InStock
TCA2000-5G
TCA2000-25G
5-Aminonicotinic acid ≥98.0% (by HPLC, titration analysis)
5-Aminonicotinic acid
CAS Number: 24242-19-1
MDL Number: MFCD00129116
Molecular Formula: C6H6N2O2
Molecular Weight: 138.13
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 293
Other Product Names / Synonyms: 5-Aminopyridine-3-carboxylic Acid