Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3'-Chloroacetophenone ≥97.0%

Supplier: TCI America
Synonyms: 1-(3-Chlorophenyl)ethanonem-Chloroacetophenone

C0096-25ML C0096-10ML
TCC0096-010MLEA 36.54 USD InStock
TCC0096-010ML TCC0096-25ML
3'-Chloroacetophenone ≥97.0%
3'-Chloroacetophenone
CAS Number: 99-02-5
MDL Number: MFCD00000593
Molecular Formula: C8H7ClO
Molecular Weight: 154.59
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 230
Flash Point (°C): 105
Specific Gravity (20/20): 1.20


Formula: C₈H₇ClO
MW: 154.6 g/mol
Boiling Pt: 230 °C
Density: 1.20 (20 °C)
Flash Pt: 105 °C
MDL Number: MFCD00000593
CAS Number: 99-02-5
UN: 3416
ADR: 6.1,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/71/75/28937175.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty