Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chlorobenzylamine ≥98.0%

Supplier: TCI America
Warning
Synonyms: (4-Chlorophenyl)methanamine

C1090-25G
TCC1090-025GEA 73.08 USD InStock
TCC1090-025G
4-Chlorobenzylamine ≥98.0%
4-Chlorobenzylamine
CAS Number: 104-86-9
MDL Number: MFCD00008121
Molecular Formula: C7H8ClN
Molecular Weight: 141.60
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 214
Flash Point (°C): 90
Specific Gravity (20/20): 1.16


Formula: C₇H₈ClN
MW: 141.6 g/mol
Boiling Pt: 214 °C
Density: 1.16 (20 °C)
Flash Pt: 90 °C
MDL Number: MFCD00008121
CAS Number: 104-86-9
EINECS: 203-245-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/83/53/28938353.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty