C1693-5G
TCC1693-5GEA
42
USD
InStock
TCC1693-5G
2-Chloro-5-fluoroaniline ≥98.0%
2-Chloro-5-fluoroaniline
CAS Number: 452-83-5
MDL Number: MFCD00084852
Molecular Formula: C6H5ClFN
Molecular Weight: 145.56
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Freezing point (°C): 19
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 5-Fluoro-2-chloroaniline