Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl yellow

Supplier: TCI America
Danger
Synonyms: Dimethyl yellowSudan Yellow 4-(Dimethylamino)azobenzene 4-(N,N-Dimethylamino)azobenzene

D0231-25G
TCD0231-025GEA 30.45 USD InStock
TCD0231-025G
Methyl yellow
Methyl Yellow
CAS Number: 60-11-7
MDL Number: MFCD00008308
Molecular Formula: C14H15N3
Molecular Weight: 225.30
Form: Crystal
Color: Reddish Yellow
Melting point (°C): 114
Lambda max.: 513 nm (pH2.4)

Other Product Names / Synonyms: Butter Yellow, 4-(Dimethylamino)azobenzene, N,N-Dimethyl-4-(phenylazo)aniline, Dimethyl Yellow, Solvent Yellow 2


Formula: C₁₄H₁₅N₃
MW: 225.29 g/mol
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 114 °C
Density: 1.2233 g/cm³ (20 °C)
MDL Number: MFCD00008308
CAS Number: 60-11-7
EINECS: 200-455-7
UN: 2811
ADR: 6.1,III
Merck Index: 12,03279
https://us-legacy.vwr.com/stibo/web/std.lang.all/46/63/28974663.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty