Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4-Dinitroaniline ≥99.0%

Supplier: TCI America
Danger

D2703-25G D2703-500G
TCD2703-500GEA 751.1 USD InStock
TCD2703-500G TCD2703-025G
2,4-Dinitroaniline ≥99.0%
2,4-Dinitroaniline
CAS Number: 97-02-9
MDL Number: MFCD00007151
Molecular Formula: C6H5N3O4
Molecular Weight: 183.12
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Melting point (°C): 180
Flash Point (°C): 224


Formula: C₆H₅N₃O₄
MW: 183.12 g/mol
Boiling Pt: 401 °C (1013 hPa)
Melting Pt: 180 °C
Density: 1.6 g/cm³ (20 °C)
Flash Pt: 224 °C
Storage Temperature: Ambient
MDL Number: MFCD00007151
CAS Number: 97-02-9
EINECS: 202-553-5
UN: 1596
ADR: 6.1,II
Merck Index: 13,03298
https://us-legacy.vwr.com/stibo/web/std.lang.all/98/10/28979810.jpg
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty