Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Iodohexadecane ≥97.0%

Supplier: TCI America

I0408-25G
TCI0408-025GEA 93.38 USD InStock
TCI0408-025G
1-Iodohexadecane ≥97.0%
1-Iodohexadecane
CAS Number: 544-77-4
MDL Number: MFCD00001089
Molecular Formula: C16H33I
Molecular Weight: 352.34
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Boiling point (°C): 207
Flash Point (°C): 110
Freezing point (°C): 22

Other Product Names / Synonyms: Cetyl Iodide, Hexadecyl Iodide


Formula: C₁₆H₃₃I
MW: 352.34 g/mol
Boiling Pt: 207 °C
Melting Pt: 20…23 °C
Density: 1.12 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001089
CAS Number: 544-77-4
EINECS: 208-879-4
UN: 3082
ADR: 9,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/22/49/28972249.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty