Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(Iodomethyl)trimethylsilane ≥97.0%

Supplier: TCI America

I0449-10G
TCI0449-010GEA 92.37 USD InStock
TCI0449-010G
(Iodomethyl)trimethylsilane ≥97.0%
(Iodomethyl)trimethylsilane
CAS Number: 4206-67-1
MDL Number: MFCD00001077
Molecular Formula: C4H11ISi
Molecular Weight: 214.12
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 142
Flash Point (°C): 29
Specific Gravity (20/20): 1.44

Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: (Trimethylsilyl)methyl Iodide


Formula: C₄H₁₁ISi
MW: 214.12 g/mol
Boiling Pt: 142 °C
Density: 1.44 (20 °C)
Flash Pt: 29 °C
MDL Number: MFCD00001077
CAS Number: 4206-67-1
EINECS: 224-123-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/22/99/28972299.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty