Synonyms:
1-Aminonaphthalene
N0052-100G
N0052-25G
TCN0052-025GEA
83.59
USD
InStock
TCN0052-025G
TCN0052-100G
1-Naphthylamine ≥99.0% (by GC, titration analysis)
1-Naphthylamine
CAS Number: 134-32-7
MDL Number: MFCD00004016
Molecular Formula: C10H9N
Molecular Weight: 143.19
Purity/Analysis Method: 99.0% (GC,T)
Form: Crystal
Boiling point (°C): 301
Melting point (°C): 50
Flash Point (°C): 157
Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Bioscience, ROS/RNS Detection Reagents, Reagents for Nitric Oxide Research
Other Product Names / Synonyms: 1-Aminonaphthalene