Synonyms:
4-Nitrophenethyl alcohol
, p-Nitrophenethyl Alcohol
N0569-5G
N0569-25G
TCN0569-025GEA
184.73
USD
InStock
TCN0569-025G
TCN0569-5G
2-(4-Nitrophenyl)ethanol ≥98.0%
2-(4-Nitrophenyl)ethanol
CAS Number: 100-27-6
MDL Number: MFCD00010202
Molecular Formula: C8H9NO3
Molecular Weight: 167.16
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 148
Melting point (°C): 62
Other Product Names / Synonyms: 4-Nitrophenethyl Alcohol, 2-(4-Nitrophenyl)ethyl Alcohol