Synonyms:
Hexadecanoic acid Hexadecylic acid
P1145-5G
TCP1145-005GEA
157.39
USD
InStock
TCP1145-005G
Palmitic acid ≥99.5%
Palmitic acid
CAS Number: 57-10-3
MDL Number: MFCD00002747
Molecular Formula: C16H32O2
Molecular Weight: 256.43
Purity/Analysis Method: 99.5% (GC)
Form: Crystal
Color: White
Boiling point (°C): 352
Melting point (°C): 64
Flash Point (°C): 171
Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement, Bioscience, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Hexadecanoic Acid